Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) 96%
Leverantör: Thermo Scientific
Varning
Synonyms: Diethyl [(ethoxycarbonyl)(fluoro)methyl]phosphonate, Triethyl 2-fluoro-2-phosphonoacetate
L17913.03 L17913.06 L17913.14
L17913.03EA 1100 SEK
L17913.03 L17913.06 L17913.14
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent) 96%
Triethyl-2-fluoro-2-phosphonoacetate (fluorinated horner reagent)
Purity: 96% CAS No.: 2356-16-3 Molecular Formula: C8H16FO5P Formula Weight: 242.19 Flash Point: 73°(163°F) Refractive Index: 1.4250 Boiling Point: 75°/0.01mm Beilstein Reference: 1912161 MDL No.: MFCD00134400
Formel: C₈H₁₆FO₅P
MW: 242,19 g/mol
Kokpunkt: 75 °C (0,01 mmHg)
Densitet: 1,194
Flampunkt: 73 °C (163 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00134400
CAS nummer: 2356-16-3