Dimethyl (2-oxopropyl)phosphonate 95%
Leverantör: Thermo Scientific
Synonyms: Dimethyl acetylmethylphosphonate
177610050 177610250 177611000
ACRO177610050EA 1675 SEK
ACRO177610050 ACRO177610250 ACRO177611000
Dimethyl (2-oxopropyl)phosphonate 95%
Dimethyl (2-oxopropyl)phosphonate
Formel: C₅H₁₁O₄P
MW: 166,12 g/mol
Kokpunkt: 76…79 °C (3 mmHg)
Flampunkt: 93 °C
Storage Temperature: Omgivande
MDL Number: MFCD00008769
CAS nummer: 4202-14-6
EINECS: 224-110-5
Order Now
Specifikation testresultat
Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.4380 to 1.4410 (20°C, 589 nm)
GC ≥94 %