Diethyl (2-oxopropyl)phosphonate ≥97%
Leverantör: Thermo Fisher Scientific
Synonyms: Diethyl acetylmethylphosphonate
H26896.14 H26896.06
H26896.14EA 3875 SEK
H26896.14 H26896.06
Diethyl (2-oxopropyl)phosphonate ≥97%
Diethyl (2-oxopropyl)phosphonate
Purity: 97% CAS No.: 1067-71-6 Molecular Formula: C7H15O4P Formula Weight: 194.17 Flash Point: 113°(235°F) Refractive Index: 1.433 Boiling Point: 129°/9mm MDL No.: MFCD00044728
Formel: C₇H₁₅O₄P
Kokpunkt: 129 °C (9 mmHg)
Densitet: 1,01
Flampunkt: 113 °C (235 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00044728
CAS nummer: 1067-71-6