2-Hydroxy-1,4-naphthoquinone 99%
Leverantör: Thermo Scientific
Varning
Synonyms: 2-Hydroxy-p-naphthoquinone, 2-Hydroxy-1,4-naphoquinone
121630250 121631000 121630100
ACRO121630250EA 1050 SEK
ACRO121630250 ACRO121631000 ACRO121630100
2-Hydroxy-1,4-naphthoquinone 99%
2-Hydroxy-1,4-naphthoquinone
Formel: C₁₀H₆O₃
MW: 174,16 g/mol
Smältpunkt: 192…195 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00001678
CAS nummer: 83-72-7
EINECS: 201-496-3
Order Now
Specifikation testresultat
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥98.5 %
GC ≥98.5 %