Diethyl (4-methylbenzyl)phosphonate ≥98.0%
Leverantör: TCI
Synonyms:
Diethyl 4-methylbenzylphosphonate
D3336-25G
D3336-5G
TCIAD3336-25GEA
1200
SEK
TCIAD3336-25G
TCIAD3336-5G
Diethyl (4-methylbenzyl)phosphonate ≥98.0%
Diethyl (4-methylbenzyl)phosphonate
Formel:
C₁₂H₁₉O₃P Kokpunkt: 160°C |
MDL Number:
MFCD00041385 CAS nummer: 3762-25-2 EINECS: 223-180-4 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...