5-Bromo-1,3-benzodioxole ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: 4-Bromo-1,2-(methylenedioxy)benzene, 5-Bromobenzo[d][1,3]dioxole
A17906.06 A17906.14 A17906.22 A17906.36
A17906.06EA 665 SEK
A17906.06 A17906.14 A17906.22 A17906.36
5-Bromo-1,3-benzodioxole ≥98%
5-Bromo-1,3-benzodioxole
Purity: 98% CAS No.: 2635-13-4 Molecular Formula: C7H5BrO2 Formula Weight: 201.02 Flash Point: 110°(230°F) Refractive Index: 1.5840 Boiling Point: 228° Beilstein Reference: 127407 MDL No.: MFCD00005821
Formel: C₇H₅BrO₂
MW: 201,02 g/mol
Kokpunkt: 228 °C
Densitet: 1,669
Flampunkt: >110 °C (230 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00005821
CAS nummer: 2635-13-4
EINECS: 220-123-5