Triethyl-2-phosphonopropionate 98%
Leverantör: Thermo Scientific
Varning
Synonyms: Diethyl [1-(ethoxycarbonyl)ethyl]phosphonate, Ethyl 2-(diethoxyphosphoryl)propanoate
174360100 174360500
ACRO174360100EA 985 SEK
ACRO174360100 ACRO174360500
Triethyl-2-phosphonopropionate 98%
Triethyl-2-phosphonopropionate
Formel: C₉H₁₉O₅P
MW: 238,22 g/mol
Kokpunkt: 143…144 °C (12 mmHg)
Densitet: 1,11 g/cm³
Flampunkt: 88 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00009159
CAS nummer: 3699-66-9
EINECS: 223-033-4
Order Now
Specifikation testresultat
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4310 to 1.4330 (20°C, 589 nm)
GC ≥97.5 %