2-Propyl-2-oxazoline ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
2-Propyl-4,5-dihydrooxazole
P1739-1G
P1739-5G
TCIAP1739-1GEA
615
SEK
TCIAP1739-1G
TCIAP1739-5G
2-Propyl-2-oxazoline ≥98.0%
2-Propyl-2-oxazoline
Formel:
C₆H₁₁NO MW: 113,157 g/mol Kokpunkt: 147°C |
MDL Number:
MFCD02684776 CAS nummer: 4694-80-8 EINECS: 225-156-9 UN: 1993 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...