2-Chlorophenyl phosphorodichloridate 98+%, stavar
Leverantör: Thermo Fisher Scientific
Fara
A11505.06 A11505.14 A11505.22
A11505.06EA 595 SEK
A11505.06 A11505.14 A11505.22
2-Chlorophenyl phosphorodichloridate 98+%, stavar
2-Chlorophenyl phosphorodichloridate
Purity: 98+% CAS No.: 15074-54-1 Molecular Formula: C6H4Cl3O2P Formula Weight: 245.43 Flash Point: 105°(221°F) Refractive Index: 1.5380 Boiling Point: 82-84°/0.4mm Beilstein Reference: 911009 UN No.: 3265 MDL No.: MFCD00009704
Formel: C₆H₄Cl₃O₂P
MW: 245.43 g/mol
Kokpunkt: 82…84 °C (0.4 mmHg)
Densitet: 1.491
Flampunkt: 105 °C (221 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009704
CAS nummer: 15074-54-1
EINECS: 239-129-4
UN: 3265
ADR: 8 II