α-Chloropropiophenone ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Chloro-1-phenyl-1-propanone, 2-Chloropropiophenone

C1647-25G C1647-5G
TCIAC1647-25GEA 3450 SEK
TCIAC1647-25G TCIAC1647-5G
α-Chloropropiophenone ≥95.0%
α-Chloropropiophenone
Formel: C₉H₉ClO
MDL Number: MFCD03844782
CAS nummer: 6084-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4741c825e169780e2b16826db4beb81e55415519&vid=4741c825e169780e2b16826db4beb81e55415519&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR