Ethyl-4,4,4-trifluorocrotonate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Ethyl (E)-4,4,4-trifluorobut-2-enoate, Ethyl 4,4,4-trifluorocrotonate
E0772-25G E0772-5G
TCIAE0772-25GEA 3325 SEK
TCIAE0772-25G TCIAE0772-5G
Ethyl-4,4,4-trifluorocrotonate ≥98.0%
Ethyl-4,4,4-trifluorocrotonate
Formel: C₆H₇F₃O₂
MW: 168,12 g/mol
Kokpunkt: 115°C
Flampunkt: 24°C
MDL Number: MFCD00009903
CAS nummer: 25597-16-4
UN: 3272
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d45b4df36069f1f0b6431bbb490289f04921ee1b&vid=d45b4df36069f1f0b6431bbb490289f04921ee1b&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %