1-Naphthoyl chloride 99%
Leverantör: Thermo Scientific
Fara
270380100 270380500
ACRO270380100EA 800 SEK
ACRO270380100 ACRO270380500
1-Naphthoyl chloride 99%
1-Naphthoyl chloride
Formel: C₁₁H₇ClO
MW: 190,63 g/mol
Kokpunkt: 190 °C (35 mmHg)
Smältpunkt: 16…19 °C
Densitet: 1,26 g/cm³
Flampunkt: >110 °C
Storage Temperature: Frys
MDL Number: MFCD00004002
CAS nummer: 879-18-5
EINECS: 212-903-9
UN: 3265
ADR: 8,II
Order Now
Specifikation testresultat
Appearance Clear yellow to beige to orange-brown Liquid
Infrared spectrum Conforms
Refractive index 1.6510 to 1.6540 (20°C, 589 nm)
GC ≥98.5 %