4,4'-Dihydroxydiphenylmethane ≥99.0%

Leverantör: TCI
Varning

B0819.0025 B0819-25G B0819-5G B0819-100G
TCIAB0819.0025EA 2625 SEK
TCIAB0819.0025 TCIAB0819-25G TCIAB0819-5G TCIAB0819-100G
4,4'-Dihydroxydiphenylmethane ≥99.0%
4,4'-Dihydroxydiphenylmethane
Formel: C₁₃H₁₂O₂
MDL Number: MFCD00002385
CAS nummer: 620-92-8
EINECS: 210-658-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4c52082483169cd7c1b540c876d9d30d211cef7&vid=f4c52082483169cd7c1b540c876d9d30d211cef7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 99.0 %
Melting point 160.0 to 164.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR