2,2'-Dihydroxydiphenylmethane ≥99.0%
Leverantör: TCI
|
Varning
|
D1939-5G
D1939-1G
TCIAD1939-5GEA
2975
SEK
TCIAD1939-5G
TCIAD1939-1G
2,2'-Dihydroxydiphenylmethane ≥99.0%
2,2'-Dihydroxydiphenylmethane
Formel:
C₁₃H₁₂O₂ |
MDL Number:
MFCD00002243 CAS nummer: 2467-02-9 EINECS: 219-578-2 |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 99.0 % |
Melting point | 117.0 to 120.0 deg-C |
Solubility in Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...