2,4-Dichlorothiazole ≥98.0%
Leverantör: TCI EUROPE
Varning
D3549-25G D3549-5G
TCIAD3549-25GEA 3625 SEK
TCIAD3549-25G TCIAD3549-5G
2,4-Dichlorothiazole ≥98.0%
2,4-Dichlorothiazole
Formel: C₃HCl₂NS
MW: 154,0200 g/mol
Kokpunkt: 184°C
MDL Number: MFCD08691358
CAS nummer: 4175-76-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8037328004592bd3a59453a0355292025076c78&vid=a8037328004592bd3a59453a0355292025076c78&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C