4,4'-Dibromooctafluorobiphenyl (DBOB) ≥95.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4,4-Dibromooctafluorobiphenyl
D1696-1G D1696-5G
TCIAD1696-1GEA 765 SEK
TCIAD1696-1G TCIAD1696-5G
4,4'-Dibromooctafluorobiphenyl (DBOB) ≥95.0%
4,4'-Dibromooctafluorobiphenyl (DBOB)
Formel: C₁₂Br₂F₈
MW: 455,94 g/mol
MDL Number: MFCD00000310
CAS nummer: 10386-84-2
EINECS: 233-847-1
UN: 3152
ADR: 9,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c210eaa46cf493960e29848bfca87724927fec2e&vid=c210eaa46cf493960e29848bfca87724927fec2e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 113.0 to 116.0 deg-C