(S)-(+)-Nipecotic acid ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: L-Nipecotic acid, (S)-(+)-3-Piperidinecarboxylic acid, (S)-Piperidine-3-carboxylic acid

N0678-1G N0678-5G
TCIAN0678-1GEA 295 SEK
TCIAN0678-1G TCIAN0678-5G
(S)-(+)-Nipecotic acid ≥98.0%
(S)-(+)-Nipecotic acid
Formel: C₆H₁₁NO₂
MW: 129,16 g/mol
MDL Number: MFCD01630807
CAS nummer: 59045-82-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2845568f0faa85aa09032a56cbe4d5d9f2c7e94&vid=a2845568f0faa85aa09032a56cbe4d5d9f2c7e94&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White powder to crystal
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D +5.0 to +6.0 deg (C=5, H2O)
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR