Methyl-3-hydroxy-2-naphthoate ≥98%
Leverantör: Thermo Scientific
Varning
Synonyms: Methyl 3-hydroxy-2-naphthoate
L14533.14 L14533.06
L14533.14EA 2675 SEK
L14533.14 L14533.06
Methyl-3-hydroxy-2-naphthoate ≥98%
Methyl-3-hydroxy-2-naphthoate
Purity: 98+% CAS No.: 883-99-8 Molecular Formula: C12H10O3 Formula Weight: 202.21 Melting Point: 72-74° Boiling Point: 205-207°/160mm MDL No.: MFCD00004099
Formel: C₁₂H₁₀O₃
MW: 202,21 g/mol
Kokpunkt: 205…207 °C (160 mmHg)
Smältpunkt: 72…74 °C
Storage Temperature: Omgivande
MDL Number: MFCD00004099
CAS nummer: 883-99-8
EINECS: 212-936-9