N,N-Diethylcyclohexylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI
|
Fara
|
D0484-25ML
D0484-500ML
TCIAD0484-25MLEA
230
SEK
TCIAD0484-25ML
TCIAD0484-500ML
N,N-Diethylcyclohexylamine ≥98.0% (by GC, titration analysis)
N,N-Diethylcyclohexylamine
Formel:
C₁₀H₂₁N Kokpunkt: 193°C Flampunkt: 57°C |
MDL Number:
MFCD00003847 CAS nummer: 91-65-6 EINECS: 202-087-2 UN: 2734 ADR: 8,II |
|
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...