Methyl coumalate ≥96.0%
Leverantör: TCI EUROPE
Synonyms:
Methyl 2-oxo-2H-pyran-5-carboxylate
C1734-25G
C1734-5G
TCIAC1734-25GEA
2275
SEK
TCIAC1734-25G
TCIAC1734-5G
Methyl coumalate ≥96.0%
Methyl coumalate
Formel:
C₇H₆O₄ MW: 154,12 g/mol Kokpunkt: 260°C |
MDL Number:
MFCD00010120 CAS nummer: 6018-41-3 EINECS: 227-871-1 Merck Index: 14,02557 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 67.0 to 73.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...