(R)-(+)-α,α-Diphenylprolinol ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
D2365-1G
D2365-5G
TCIAD2365-1GEA
1025
SEK
TCIAD2365-1G
TCIAD2365-5G
(R)-(+)-α,α-Diphenylprolinol ≥98.0% (by GC, titration analysis)
(R)-(+)-α,α-Diphenylprolinol
Formel:
C₁₇H₁₉NO MW: 253,35 g/mol |
MDL Number:
MFCD00077754 CAS nummer: 22348-32-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Optical purity (LC) | min. 99.5 ee% |
Melting point | 77.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...