Tris[2-(2-methoxyethoxy)ethyl]amine 90,0 GC_ANALYSMETOD
Leverantör: TCI
|
Fara
|
Synonyms:
Tris(2-(2-methoxyethoxy)ethyl)amine, Tris(3,6-dioxaheptyl)amine, Tris(dioxa-3,6-heptyl)amine
T1231-25G
T1231-500G
T1231-100G
TCIAT1231-25GEA
350
SEK
TCIAT1231-25G
TCIAT1231-500G
TCIAT1231-100G
Tris[2-(2-methoxyethoxy)ethyl]amine 90,0 GC_ANALYSMETOD
Tris[2-(2-methoxyethoxy)ethyl]amine
Formel:
C₁₅H₃₃NO₆ MW: 323.43 g/mol Kokpunkt: 171°C Flampunkt: 162°C |
MDL Number:
MFCD00010748 CAS nummer: 70384-51-9 EINECS: 274-590-5 UN: 2735 ADR: 8 III |
|
Specifikation testresultat
Appearance | Light yellow to Amber to Dark green clear liquid |
Purity (GC) | min. 90.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...