3,5-Dichloro-2-pyridone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3,5-Dichloro-2-hydroxypyridine
D3312-25G
D3312-5G
TCIAD3312-25GEA
3650
SEK
TCIAD3312-25G
TCIAD3312-5G
3,5-Dichloro-2-pyridone ≥98.0%
3,5-Dichloro-2-pyridone
Formel:
C₅H₃Cl₂NO MW: 163,99 g/mol |
MDL Number:
MFCD00015401 CAS nummer: 5437-33-2 EINECS: 226-609-3 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 178.0 to 182.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...