1,5-Dioxepan-2-one ≥98.0%
Leverantör: TCI
D3707-25G D3707-5G
TCIAD3707-25GEA 10950 SEK
TCIAD3707-25G TCIAD3707-5G
1,5-Dioxepan-2-one ≥98.0%
1,5-Dioxepan-2-one
Formel: C₅H₈O₃
Storage Temperature: Frys
CAS nummer: 35438-57-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a64c2a8ad5b6118695a370c18a2809a914ca53f7&vid=a64c2a8ad5b6118695a370c18a2809a914ca53f7&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 34.0 to 38.0 deg-C