(±)-3-Hydroxytetrahydrofuran ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Hydroxytetrahydrofuran, Oxolan-3-ol
H1331-25G
H1331-5G
TCIAH1331-25GEA
1475
SEK
TCIAH1331-25G
TCIAH1331-5G
(±)-3-Hydroxytetrahydrofuran ≥97.0%
(±)-3-Hydroxytetrahydrofuran
Formel:
C₄H₈O₂ MW: 88,1100 g/mol Kokpunkt: 181°C Flampunkt: 81°C |
MDL Number:
MFCD00005374 CAS nummer: 453-20-3 EINECS: 207-219-2 |
Specifikation testresultat
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...