1,4,7-Triazacyclononane ≥98.0%

Leverantör: TCI EUROPE
Fara

Synonyms: 1,4,7-Triazonane, TACN

T1878-1G T1878-200MG T1878-5G
TCIAT1878-1GEA 2500 SEK
TCIAT1878-1G TCIAT1878-200MG TCIAT1878-5G
1,4,7-Triazacyclononane ≥98.0%
1,4,7-Triazacyclononane
Formel: C₆H₁₅N₃
MW: 129,2 g/mol
Kokpunkt: 113°C
Smältpunkt: 45°C
MDL Number: MFCD00012358
CAS nummer: 4730-54-5
UN: 3259
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f18bf6fa8f0af36b39425415d8d6c9d6e1e4b5e&vid=8f18bf6fa8f0af36b39425415d8d6c9d6e1e4b5e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR