meso-1,2,3,4-Tetrabromobutane ≥98.0%
Leverantör: TCI EUROPE
T0775-25G T0775-500G
TCIAT0775-25GEA 405 SEK
TCIAT0775-25G TCIAT0775-500G
meso-1,2,3,4-Tetrabromobutane ≥98.0%
meso-1,2,3,4-Tetrabromobutane
Formel: BrH₂C(BrCH)₂CH₂Br
Kokpunkt: 181°C
MDL Number: MFCD00070471
CAS nummer: 2657-67-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=412831859fb4c2fc34d3c9b666d7649493a1cff8&vid=412831859fb4c2fc34d3c9b666d7649493a1cff8&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 119.0 deg-C
Solubility in hot Methanol almost transparency