O-(4-Chlorophenyl) chlorothioformate ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: Chlorothioformic acid O-(4-Chlorophenyl) ester, 4-Chlorophenyl chlorothioformate, 4-Chlorophenyl chlorothionoformate
C1588-25G C1588-5G
TCIAC1588-25GEA 3675 SEK
TCIAC1588-25G TCIAC1588-5G
O-(4-Chlorophenyl) chlorothioformate ≥98.0%
O-(4-Chlorophenyl) chlorothioformate
Formel: C₇H₄Cl₂OS
Kokpunkt: 131°C
MDL Number: MFCD00014465
CAS nummer: 937-64-4
EINECS: 213-334-9
UN: 3265
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=079b57bded29cce84d0ae8a95aeb6c39c87b39d6&vid=079b57bded29cce84d0ae8a95aeb6c39c87b39d6&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Light yellow to Amber to Dark green clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 98.0 %