1,3-Phenylene diisocyanate ≥98.0%

Leverantör: TCI EUROPE
Fara

P1092-1G P1092-5G
TCIAP1092-1GEA 590 SEK
TCIAP1092-1G TCIAP1092-5G
1,3-Phenylene diisocyanate ≥98.0%
1,3-Phenylene diisocyanate
Formel: C₈H₄N₂O₂
Kokpunkt: 103°C
Storage Temperature: Kylskåp
MDL Number: MFCD00002018
CAS nummer: 123-61-5
EINECS: 204-637-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1631ca77ce16df05da9d2fd4ca538eccfd11c3b&vid=a1631ca77ce16df05da9d2fd4ca538eccfd11c3b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 49.0 to 54.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR