Methyl-3-(methylthio)propionate
Leverantör: TCI EUROPE
Synonyms: Methyl3-(methylthio)propionate
M0862-25G M0862-500G
TCIAM0862-25GEA 270 SEK
TCIAM0862-25G TCIAM0862-500G
Methyl-3-(methylthio)propionate
Methyl-3-(methylthio)propionate
Formel: C₅H₁₀O₂S
MW: 134,20 g/mol
Kokpunkt: 81°C
Flampunkt: 72°C
MDL Number: MFCD00008565
CAS nummer: 13532-18-8
EINECS: 236-883-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c2f920b65583582c9b16d8b7789f61d8beb113e&vid=9c2f920b65583582c9b16d8b7789f61d8beb113e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 1.0