p-Xylenol blue ≥95.0%

Leverantör: TCI
Varning

Synonyms: Xylenol Blue

X0016-1G X0016-25G
TCIAX0016-1GEA 245 SEK
TCIAX0016-1G TCIAX0016-25G
p-Xylenol blue ≥95.0%
p-Xylenol blue
Formel: C₂₃H₂₂O₅S
MDL Number: MFCD00005870
CAS nummer: 125-31-5
EINECS: 204-736-5
Merck Index: 14,10083
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f7d4a34b92c24f36c0b82fda474245fee81a6dd&vid=5f7d4a34b92c24f36c0b82fda474245fee81a6dd&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Orange to Brown to Dark purple powder to crystal
Purity (HPLC) min. 95.0 area%
Lambda max. 433.0 to 437.0 nm (pH7.0)
Absorbance min. 0.370 (10 mg/L, pH 7.0, 433.0 to 437.0 nm)
Transition interval (pH) (red)1.2 to 2.8 (yellow), (yellow)8.0 to 9.6 (purplish blue)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR