Methyl-β-D-glucopyranoside hemihydrate ≥98.0%
Leverantör: TCI
M0709-25G
M0709-5G
TCIAM0709-25GEA
3775
SEK
TCIAM0709-25G
TCIAM0709-5G
Methyl-β-D-glucopyranoside hemihydrate ≥98.0%
Methyl-β-D-glucopyranoside hemihydrate
Formel:
C₇H₁₄O₆·0.5H₂O |
MDL Number:
MFCD00149442 CAS nummer: 7000-27-3 EINECS: 211-909-9 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 109.0 to 112.0 deg-C |
Specific rotation [a]20/D | -31.0 to -35.0 deg (C=5, H2O) |
Water | 4.0 to 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...