2-Methyl-3-propylpyrazine ≥95.0%

Leverantör: TCI
Varning

M0841-25ML M0841-5ML
TCIAM0841-25MLEA 1450 SEK
TCIAM0841-25ML TCIAM0841-5ML
2-Methyl-3-propylpyrazine ≥95.0%
2-Methyl-3-propylpyrazine
Formel: C₈H₁₂N₂
Flampunkt: 69°C
MDL Number: MFCD00010096
CAS nummer: 15986-80-8
EINECS: 240-121-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d3cd8ac9b84452780eba3127e73283d911df860a&vid=d3cd8ac9b84452780eba3127e73283d911df860a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR