N-Ethylpropylamine ≥97.0%

Leverantör: TCI EUROPE
Fara

E0351-25ML E0351-250ML
TCIAE0351-25MLEA 1425 SEK
TCIAE0351-25ML TCIAE0351-250ML
N-Ethylpropylamine ≥97.0%
N-Ethylpropylamine
Formel: C₅H₁₃N
MW: 87,16 g/mol
Kokpunkt: 79°C
Flampunkt: 4°C
MDL Number: MFCD00015198
CAS nummer: 20193-20-8
EINECS: 243-573-4
UN: 2733
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ea82e8f96a78a7e503d58df1edea0d8afe038efa&vid=ea82e8f96a78a7e503d58df1edea0d8afe038efa&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR