Phthalazine ≥98.0% (by GC, titration analysis)

Leverantör: TCI

Synonyms: Benzo[d]pyridazine

P0694-25G P0694-5G
TCIAP0694-25GEA 1675 SEK
TCIAP0694-25G TCIAP0694-5G
Phthalazine ≥98.0% (by GC, titration analysis)
Phthalazine
Formel: C₈H₆N₂
Kokpunkt: 317°C
MDL Number: MFCD00006908
CAS nummer: 253-52-1
EINECS: 205-963-2
Merck Index: 14,07370
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65539f9e111a1ee2cdd82f754fb28a87a14f5003&vid=65539f9e111a1ee2cdd82f754fb28a87a14f5003&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 90.0 to 93.0 deg-C
Solubility in Methanol very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR