4-Methoxycinnamaldehyde ≥97.0%

Leverantör: TCI EUROPE

Synonyms: 3-(4-Methoxyphenyl)-2-propenal, 3-(4-Methoxyphenyl)acrylaldehyde

M1012-25G M1012-5G
TCIAM1012-25GEA 990 SEK
TCIAM1012-25G TCIAM1012-5G
4-Methoxycinnamaldehyde ≥97.0%
4-Methoxycinnamaldehyde
Formel: C₁₀H₁₀O₂
MW: 162,1852 g/mol
Kokpunkt: 176°C
MDL Number: MFCD00017343
CAS nummer: 1963-36-6
EINECS: 217-807-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e45f8c011f49afae684fcbc13345ad64509f4930&vid=e45f8c011f49afae684fcbc13345ad64509f4930&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 57.0 to 61.0 deg-C
Solubility in Methanol very faint turbidity

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR