2-(2-Nitroethoxy)tetrahydropyran ≥85.0%

Leverantör: TCI
Varning

N0602-25G N0602-5G
TCIAN0602-25GEA 4300 SEK
TCIAN0602-25G TCIAN0602-5G
2-(2-Nitroethoxy)tetrahydropyran ≥85.0%
2-(2-Nitroethoxy)tetrahydropyran
Formel: C₇H₁₃NO₄
Flampunkt: 48°C
Storage Temperature: Frys
MDL Number: MFCD00010858
CAS nummer: 75233-61-3
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b14f06cfe9cef3dadcfa40bee2f1c04e7d673fec&vid=b14f06cfe9cef3dadcfa40bee2f1c04e7d673fec&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 85.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR