Acetoxyacetyl chloride ≥95.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Fara
Synonyms: Acetoxyacetylchloride, 2-Chloro-2-oxoethyl acetate
A1500-100G A1500-25G
TCIAA1500-100GEA 4275 SEK
TCIAA1500-100G TCIAA1500-25G
Acetoxyacetyl chloride ≥95.0% (by GC, titration analysis)
Acetoxyacetyl chloride
Formel: C₄H₅ClO₃
MW: 136,53 g/mol
Kokpunkt: 160°C
Flampunkt: 71°C
MDL Number: MFCD00011535
CAS nummer: 13831-31-7
EINECS: 237-542-4
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4ac28f166d5cd3f7bce2cc9f502c236c6ad8b25&vid=f4ac28f166d5cd3f7bce2cc9f502c236c6ad8b25&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 95.0 %