Ethosuximide ≥98.0% (by GC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 2-Ethyl-2-methylsuccinimide, 3-Ethyl-3-methylpyrrolidine-2,5-dione

E0746-25G E0746-5G
TCIAE0746-25GEA 1625 SEK
TCIAE0746-25G TCIAE0746-5G
Ethosuximide ≥98.0% (by GC, titration analysis)
Ethosuximide
Formel: C₇H₁₁NO₂
MW: 141.17 g/mol
Kokpunkt: 150°C
Storage Temperature: Kylskåp
MDL Number: MFCD00072123
CAS nummer: 77-67-8
EINECS: 201-048-7
Merck Index: 14,03748
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=db63924aafd7ef827cb1cd0dfa7779bb15d13715&vid=db63924aafd7ef827cb1cd0dfa7779bb15d13715&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 48.0 to 52.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR