N,N-Diethylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Diethylcarbamyl chloride
D0482-25G
D0482-500G
D0482-100G
TCIAD0482-25GEA
230
SEK
TCIAD0482-25G
TCIAD0482-500G
TCIAD0482-100G
N,N-Diethylcarbamoyl chloride ≥98.0% (by GC, titration analysis)
N,N-Diethylcarbamoyl chloride
Formel:
C₅H₁₀ClNO MW: 135,59 g/mol Flampunkt: 76°C |
MDL Number:
MFCD00000636 CAS nummer: 88-10-8 EINECS: 201-798-5 ADR: 8,II |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...