Ethyl-4,4,4-trichloroacetoacetate ≥97.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 4,4,4-Trichloroacetoacetic acid ethyl ester
T1285-10G T1285-25G
TCIAT1285-10GEA 1825 SEK
TCIAT1285-10G TCIAT1285-25G
Ethyl-4,4,4-trichloroacetoacetate ≥97.0%
Ethyl-4,4,4-trichloroacetoacetate
Formel: C₆H₇Cl₃O₃
Kokpunkt: 234°C
MDL Number: MFCD00043329
CAS nummer: 3702-98-5
EINECS: 223-041-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ca058aee52401379c3a0b935cbb2c6239811443&vid=1ca058aee52401379c3a0b935cbb2c6239811443&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %