1,6-Dibromoperfluorohexane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Perfluoro(1,6-dibromohexane)

D3572-1G D3572-5G
TCIAD3572-1GEA 350 SEK
TCIAD3572-1G TCIAD3572-5G
1,6-Dibromoperfluorohexane ≥98.0%
1,6-Dibromoperfluorohexane
Formel: C₆Br₂F₁₂
MW: 459,87 g/mol
Kokpunkt: 150°C
Storage Temperature: Kylskåp
MDL Number: MFCD00153110
CAS nummer: 918-22-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=201d3cba8efc5cfa33fcc7a5143fc724ceb8debc&vid=201d3cba8efc5cfa33fcc7a5143fc724ceb8debc&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR