3,3'-Metylendianilin ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
Varning
Synonyms: 3,3'-Methylenedianiline, 3,3'-Diaminodiphenylmethane
D1683-1G D1683-5G
TCIAD1683-1GEA 1125 SEK
TCIAD1683-1G TCIAD1683-5G
3,3'-Metylendianilin ≥98.0% (by GC, titration analysis)
3,3'-Metylendianilin
Formel: C₁₃H₁₄N₂
MDL Number: MFCD00014777
CAS nummer: 19471-12-6
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=aa25fb1e176f38819c5a1e0bd61937018c985f6e&vid=aa25fb1e176f38819c5a1e0bd61937018c985f6e&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 82.0 to 85.0 deg-C
Solubility in Methanol almost transparency