2,4-Dichlorobenzyl mercaptan ≥98.0%
Leverantör: TCI
|
Fara
|
Synonyms:
(2,4-Dichlorophenyl)methanethiol, 2,4-Dichlorobenzenemethanethiol
D2198-25G
D2198-5G
TCIAD2198-25GEA
4750
SEK
TCIAD2198-25G
TCIAD2198-5G
2,4-Dichlorobenzyl mercaptan ≥98.0%
2,4-Dichlorobenzyl mercaptan
Formel:
C₇H₆Cl₂S MW: 193.10 g/mol Kokpunkt: 115°C |
MDL Number:
MFCD00039650 CAS nummer: 59293-67-3 EINECS: 261-687-2 UN: 2810 ADR: 6.1 III |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...