Ethyl-4,4,4-trifluorobutyrate ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Ethyl 4,4,4-trifluorobutanoate, Ethyl 4,4,4-trifluorobutyrate
E0830-1G
E0830-5G
TCIAE0830-1GEA
285
SEK
TCIAE0830-1G
TCIAE0830-5G
Ethyl-4,4,4-trifluorobutyrate ≥98.0%
Ethyl-4,4,4-trifluorobutyrate
Formel:
C₆H₉F₃O₂ MW: 170,13 g/mol Kokpunkt: 127°C Flampunkt: 33°C |
MDL Number:
MFCD00041398 CAS nummer: 371-26-6 UN: 3272 ADR: 3,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...