(±)-1-Methyl-3-phenylpropylamine ≥98.0% (by GC, titration analysis)
Leverantör: TCI EUROPE
|
Fara
|
Synonyms:
3-Amino-1-phenylbutane, 3-Amino-1-phenyl-butane
A0999-25ML
A0999-500ML
TCIAA0999-25MLEA
510
SEK
TCIAA0999-25ML
TCIAA0999-500ML
(±)-1-Methyl-3-phenylpropylamine ≥98.0% (by GC, titration analysis)
(±)-1-Methyl-3-phenylpropylamine
Formel:
C₁₀H₁₅N MW: 149,24 g/mol Kokpunkt: 222°C Flampunkt: 97°C |
MDL Number:
MFCD00008090 CAS nummer: 22374-89-6 EINECS: 244-942-2 UN: 2735 ADR: 8,III |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...