Methyl-4-methoxyacetoacetate ≥97.0%

Leverantör: TCI
Varning

Synonyms: 4-Methoxyacetoacetic acid methyll ester, Methyl 4-methoxy-3-oxobutanoate

M1031-25G M1031-5G
TCIAM1031-25GEA 1100 SEK
TCIAM1031-25G TCIAM1031-5G
Methyl-4-methoxyacetoacetate ≥97.0%
Methyl-4-methoxyacetoacetate
Formel: C₆H₁₀O₄
Kokpunkt: 113°C
Flampunkt: 89°C
MDL Number: MFCD00010183
CAS nummer: 41051-15-4
EINECS: 255-188-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=135d6f5f74821562b0f525fab2dc2571c7d85406&vid=135d6f5f74821562b0f525fab2dc2571c7d85406&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR