1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 1,3-Bis(2,6-di-i-propylphenyl)imidazolium chloride, IPr.HCl

D3611-5G D3611-500MG D3611-25G
TCIAD3611-5GEA 2975 SEK
TCIAD3611-5G TCIAD3611-500MG TCIAD3611-25G
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride ≥98.0% (by HPLC, titration analysis)
1,3-Bis(2,6-diisopropylphenyl)imidazolium chloride
Formel: C₂₇H₃₇ClN₂
MW: 425,05 g/mol
MDL Number: MFCD02684545
CAS nummer: 250285-32-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ece60b8e5bb1008cf1c730c5eb465269031821e7&vid=ece60b8e5bb1008cf1c730c5eb465269031821e7&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR