Inosine ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
9-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one
I0037-25G
I0037-500G
TCIAI0037-25GEA
575
SEK
TCIAI0037-25G
TCIAI0037-500G
Inosine ≥98.0% (by HPLC, titration analysis)
Inosine
Formel:
C₁₀H₁₂N₄O₅ MW: 268,23 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00066770 CAS nummer: 58-63-9 EINECS: 200-390-4 Merck Index: 14,04975 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -50.0 to -53.0 deg (C=1, H2O) |
Solubility in Water | almost transparency |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...