1,2-Bis(diphenylphosphino)benzene ≥98.0%

Leverantör: TCI EUROPE

B3372-1G B3372-5G
TCIAB3372-1GEA 805 SEK
TCIAB3372-1G TCIAB3372-5G
1,2-Bis(diphenylphosphino)benzene ≥98.0%
1,2-Bis(diphenylphosphino)benzene
Formel: C₃₀H₂₄P₂
MW: 446,46 g/mol
MDL Number: MFCD00014081
CAS nummer: 13991-08-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5290b74c660a6379e5d19eefafd47c419148e9c5&vid=5290b74c660a6379e5d19eefafd47c419148e9c5&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 186.0 to 190.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR